O-Benzylhydroxylamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

A17393-22 A17393-06 A17393-14
CAAAAA17393-22EA 812.12 CAD
CAAAAA17393-22 CAAAA17393-06 CAAAA17393-14
O-Benzylhydroxylamine hydrochloride 99%
O-Benzylhydroxylamine hydrochloride

Fieser: 14,28


Purity: 99% CAS No.: 2687-43-6 Molecular Formula: C6H5CH2ONH2.HCl Formula Weight: 159.62 Melting Point: ca 238° subl. Beilstein Reference: 3595440

Formula: C₇H₁₀ClNO
MW: 159.62 g/mol
Melting Pt: ∼ 238 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00012952
CAS Number: 2687-43-6
EINECS: 220-249-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR