EDTA tetrasodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A17385-0B A17385-0I A17385-30
CAAAA17385-0BEA 112.81 CAD
CAAAA17385-0B CAAAA17385-0I CAAAA17385-30
EDTA tetrasodium salt hydrate 98%
EDTA tetrasodium salt hydrate

Purity: 98% CAS No.: 194491-31-1 Molecular Formula: C10H12N2Na4O8.xH2O Formula Weight: 380.17(anhy) Melting Point: 300° Beilstein Reference: 3828865 MDL No.: MFCD00150025

Formula: C₁₀H₁₄N₂Na₄O₉
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150025
CAS Number: 194491-31-1
EINECS: 200-573-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR