1-Benzyl-1-phenylhydrazine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17367-06
A17367-14
CAAAA17367-06EA
77.89
CAD
CAAAA17367-06
CAAAA17367-14
1-Benzyl-1-phenylhydrazine hydrochloride 98+%
1-Benzyl-1-phenylhydrazine hydrochloride
Purity: 98+%
CAS No.: 5705-15-7
Molecular Formula: C13H14N2.HCl
Formula Weight: 234.73
Melting Point: 178-181°
Beilstein Reference: 3707186
Formula:
C₁₃H₁₅ClN₂ Melting Pt: 178…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050690 CAS Number: 5705-15-7 EINECS: 227-195-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...