Ethyl-4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals

A17365-09 A17365-18 A17365-30
CAAAA17365-09EA 132.4 CAD
CAAAA17365-09 CAAAA17365-18 CAAAA17365-30
Ethyl-4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 98%
Ethyl-4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate

Purity: 98% CAS No.: 2386-26-7 Molecular Formula: C11H15NO3 Formula Weight: 209.25 Melting Point: 140-143° MDL No.: MFCD00022373

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00022373
CAS Number: 2386-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR