Amberlyst® A-26 (OH), Amberlyst™ ion exchange resin

Supplier: Thermo Scientific Chemicals
Danger

A17361-0B A17361-30
CAAAAA17361-0BEA 325.48 CAD
CAAAAA17361-0B CAAAAA17361-30
Amberlyst® A-26 (OH), Amberlyst™ ion exchange resin
Amberlyst® A-26 (OH)

Macroreticular strongly basic anion exchange resin, hydroxide form, suitable for aqueous and non-aqueous applications. Useful for removal of anionic metal complexes and mercaptans from non-polar solvents, and of acidic impurities from organic solutions.


CAS No.: 39339-85-0 MDL No.: MFCD00145579

Storage Temperature: Ambient
MDL Number: MFCD00145579
CAS Number: 39339-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR