12-Hydroxystearic acid ≥85%, tech.
Supplier: Thermo Scientific Chemicals
Synonyms:
12-Hydroxyoctadecanoic Acid
A17347-06
A17347-14
A17347-22
CAAAA17347-06EA
52.77
CAD
CAAAA17347-06
CAAAA17347-14
CAAAA17347-22
12-Hydroxystearic acid ≥85%, tech.
12-Hydroxystearic acid
Purity: 85%
CAS No.: 106-14-9
Molecular Formula: C18H36O3
Formula Weight: 300.48
Melting Point: 72-77°
Beilstein Reference: 1726730
MDL No.: MFCD00004592
Formula:
C₁₈H₃₆O₃ Melting Pt: 72…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004592 CAS Number: 106-14-9 EINECS: 203-366-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...