4-(Chlorosulfonyl)benzoic acid 96%

Supplier: Thermo Scientific Chemicals
Danger

A17337-03 A17337-06 A17337-14
CAAAA17337-03EA 47.24 CAD
CAAAA17337-03 CAAAA17337-06 CAAAA17337-14
4-(Chlorosulfonyl)benzoic acid 96%
4-(Chlorosulphonyl)benzoic acid

Purity: 96% CAS No.: 10130-89-9 Molecular Formula: C7H5ClO4S Formula Weight: 220.63 Odor: Acrid Melting Point: 228-232° UN No.: 3261 MDL No.: MFCD00007448

Formula: C₇H₅ClO₄S
MW: 220.63 g/mol
Melting Pt: 228…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00007448
CAS Number: 10130-89-9
EINECS: 233-367-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR