3-Acetyl-2,5-dimethylthiophene 99%
Supplier: Thermo Scientific Chemicals
A17329-09
A17329-18
CAAAA17329-09EA
73.94
CAD
CAAAA17329-09
CAAAA17329-18
3-Acetyl-2,5-dimethylthiophene 99%
3-Acetyl-2,5-dimethylthiophene
Purity: 99%
CAS No.: 2530-10-1
Molecular Formula: C8H10OS
Formula Weight: 154.23
Flash Point: 98°(208°F)
Refractive Index: 1.5455
Boiling Point: 224°
Beilstein Reference: 112095
MDL No.: MFCD00009763
Formula:
C₈H₁₀OS Boiling Pt: 224 °C Density: 1.086 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009763 CAS Number: 2530-10-1 EINECS: 219-779-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...