Cyclohexylamine hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17308-22
A17308-36
CAAAA17308-22EA
59.41
CAD
CAAAA17308-22
CAAAA17308-36
Cyclohexylamine hydrochloride ≥98%
Cyclohexylamine hydrochloride
Purity: 98+%
CAS No.: 4998-76-9
Molecular Formula: C6H13N.HCl
Formula Weight: 135.64
Odor: Odorless
Melting Point: 206-209°
Beilstein Reference: 3613002
MDL No.: MFCD00050540
Formula:
C₆H₁₃N Melting Pt: 206…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050540 CAS Number: 4998-76-9 EINECS: 225-661-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...