4-Nitrophenyl phenyl sulfide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (4-Nitrophenyl)(phenyl)sulphane, (4-Nitrophenyl)(phenyl)sulfane

A17278-06 A17278-14
CAAAA17278-06EA 112.97 CAD
CAAAA17278-06 CAAAA17278-14
4-Nitrophenyl phenyl sulfide 98%
4-Nitrophenyl phenyl sulphide

Purity: 98% CAS No.: 952-97-6 Molecular Formula: C12H9NO2S Formula Weight: 231.27 Melting Point: 54-57° MDL No.: MFCD00024700

Formula: C₁₂H₉NO₂S
MW: 231.27 g/mol
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00024700
CAS Number: 952-97-6
EINECS: 213-462-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR