Ethyl-3-aminopropanoate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17277-09
A17277-18
CAAAA17277-09EA
91.17
CAD
CAAAA17277-09
CAAAA17277-18
CAAAA17277-30
Ethyl-3-aminopropanoate hydrochloride 98%
Ethyl-3-aminopropanoate hydrochloride
Purity: 98%
CAS No.: 4244-84-2
Molecular Formula: H2NCH2CH2CO2CH2CH3.HCl
Formula Weight: 153.61
Melting Point: 67-72°
Beilstein Reference: 3559095
MDL No.: MFCD00012909
Formula:
C₅H₁₁NO₂·HCl Melting Pt: 67…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012909 CAS Number: 4244-84-2 EINECS: 224-203-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...