Dimethyl-2,5-pyridinedicarboxylate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl pyridine-2,5-dicarboxylate

A17250-22 A17250-06 A17250-14
CAAAAA17250-22EA 810.54 CAD
CAAAAA17250-22 CAAAA17250-03 CAAAA17250-06 CAAAA17250-14
Dimethyl-2,5-pyridinedicarboxylate 98+%
Dimethyl 2,5-Pyridinedicarboxylate

Purity: 98+% CAS No.: 881-86-7 Molecular Formula: C9H9NO4 Formula Weight: 195.17 Melting Point: 160-163° Beilstein Reference: 172251

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00034767
CAS Number: 881-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR