L-Glutamic acid monopotassium salt monohydrate ≥97%

Supplier: Thermo Scientific Chemicals

A17232-0B A17232-30
CAAAA17232-0BEA 366.56 CAD
CAAAA17232-0B CAAAA17232-30
L-Glutamic acid monopotassium salt monohydrate ≥97%
L-Glutamic acid monopotassium salt monohydrate

Purity: 97+% CAS No.: 6382-01-0 Molecular Formula: C5H8KNO4.H2O Formula Weight: 203.25 (185.23anhy) Melting Point: 115-118° MDL No.: MFCD00150343

Formula: C₅H₈KNO₄·H₂O
MW: 203.25 g/mol (185.23 anhy)
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00150343
CAS Number: 6382-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR