1-Acetamidoadamantane 98%

Supplier: Thermo Scientific Chemicals
Warning

A17224-09 A17224-18
CAAAA17224-09EA 52.77 CAD
CAAAA17224-09 CAAAA17224-18
1-Acetamidoadamantane 98%
1-Acetamidoadamantane

Purity: 98% CAS No.: 880-52-4 Molecular Formula: C12H19NO Formula Weight: 193.29 Melting Point: 146-149° MDL No.: MFCD00074730

Formula: C₁₂H₁₉NO
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00074730
CAS Number: 880-52-4
EINECS: 212-914-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR