N-tert-Butylisopropylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17221-06
A17221-14
CAAAA17221-06EA
109.97
CAD
CAAAA17221-06
CAAAA17221-14
N-tert-Butylisopropylamine 98%
N-tert-Butylisopropylamine
Purity: 98%
CAS No.: 7515-80-2
Molecular Formula: (CH3)3CNHCH(CH3)2
Formula Weight: 115.22
Flash Point: -3°(26°F)
Refractive Index: 1.3980
Boiling Point: 98°
Beilstein Reference: 1846752
UN No.: 2733
MDL No.: MFCD00037059
Formula:
(CH₃)₃CNHCH(CH₃)₂ Boiling Pt: 98 °C Density: 0.727 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037059 CAS Number: 7515-80-2 EINECS: 231-370-3 UN: 2733 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...