Menthofuran 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17192-03
A17192-06
CAAAA17192-03EA
75.21
CAD
CAAAA17192-03
CAAAA17192-06
Menthofuran 95%
Menthofuran
Purity: 95%
CAS No.: 494-90-6
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 75°(167°F)
Refractive Index: 1.4845
Boiling Point: 86-88°/15mm
MDL No.: MFCD00041851
Formula:
C₁₀H₁₄O Boiling Pt: 86…88 °C/15 mm Density: 0.972 Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041851 CAS Number: 494-90-6 EINECS: 207-795-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...