(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH
A17182-06
A17182-14
CAAAA17182-06EA
48.03
CAD
CAAAA17182-06
CAAAA17182-14
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Purity: 99%
CAS No.: 1161-13-3
Molecular Formula: C17H17NO4
Formula Weight: 299.33
Melting Point: 86-92°
Beilstein Reference: 2222826
Optical Rotation: +5° (c=5 in acetic acid)
MDL No.: MFCD00020418
Formula:
C17H17NO4 MW: 299.3 g/mol Melting Pt: 86…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020418 CAS Number: 1161-13-3 EINECS: 214-599-3 |
Specification Test Results
Optical rotation | +5° (c=5 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...