Nile blue A

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulfate, Nile blue A sulphate, Nile blue sulphate, Nile blue A sulfate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Nile blue sulfate, Basic Blue 12

A17174-06 A17174-14
CAAAA17174-06EA 48.77 CAD
CAAAA17174-06 CAAAA17174-14
Nile blue A
Nile blue A

CAS No.: 3625-57-8 Molecular Formula: C40H40N6O6S Formula Weight: 732.84 Melting Point: 300° dec. MDL No.: MFCD00064529

Formula: C₄₀H₄₀N₆O₆S
MW: 732.84 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064529
CAS Number: 3625-57-8
EINECS: 222-832-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR