N-Carbamoyl-DL-aspartic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Ureidosuccinic acid, N-Carbamyl-DL-aspartic Acid

A17166-09 A17166-18
CAAAA17166-09EA 598.82 CAD
CAAAA17166-09 CAAAA17166-18
N-Carbamoyl-DL-aspartic acid 98%
N-Carbamoyl-DL-aspartic acid

Purity: 98% CAS No.: 923-37-5 Molecular Formula: C5H8N2O5 Formula Weight: 176.13 Melting Point: ca 175° dec. Beilstein Reference: 1726861 MDL No.: MFCD00042822

Formula: C₅H₈N₂O₅
MW: 176.13 g/mol
Melting Pt: ∼ 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00042822
CAS Number: 923-37-5
EINECS: 213-096-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR