Methyl-2,1,3-benzothiadiazole-5-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2,1,3-benzothiadiazole-5-carboxylate

A17111-03 A17111-06
CAAAA17111-03EA 116.45 CAD
CAAAA17111-03 CAAAA17111-06 CAAAA17111-14
Methyl-2,1,3-benzothiadiazole-5-carboxylate 98%
Methyl-2,1,3-benzothiadiazole-5-carboxylate

Purity: 98% CAS No.: 175204-21-4 Molecular Formula: C8H6N2O2S Formula Weight: 194.21 Melting Point: 90-92° MDL No.: MFCD00173948

Formula: C₈H₆N₂O₂S
MW: 194.21 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00173948
CAS Number: 175204-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR