Xanthotoxin 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9-Methoxy-7H-furo[3,2-g]chromen-7-one, 8-Methoxypsoralen, Methoxsalen

A17108-03 A17108-06 A17108-14
CAAAA17108-03EA 70.63 CAD
CAAAA17108-03 CAAAA17108-06 CAAAA17108-14
Xanthotoxin 99%
Xanthotoxin

A naturally occuring photoreactive substance


Purity: 99% CAS No.: 298-81-7 Molecular Formula: C12H8O4 Formula Weight: 216.20 Melting Point: 146-150° Beilstein Reference: 196453 MDL No.: MFCD00005009

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005009
CAS Number: 298-81-7
EINECS: 206-066-9
UN: 3261
ADR: 8 III
Merck Index: 14,5988

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR