3-Indoxyl sulfate potassium salt 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Indolyl sulfate potassium salt, Potassium 1H-indol-3-yl sulphate, Potassium 1H-indol-3-yl sulfate, 3-Indolyl sulphate potassium salt
A17079-01
A17079-03
A17079-06
CAAAA17079-01EA
109.97
CAD
CAAAA17079-01
CAAAA17079-03
CAAAA17079-06
3-Indoxyl sulfate potassium salt 97%
3-Indoxyl sulfate potassium salt
Purity: 97%
CAS No.: 2642-37-7
Molecular Formula: C8H6KNO4S
Formula Weight: 251.31
Storage Temperature: -30°C to -10°C
Melting Point: 165° dec.
Beilstein Reference: 3921324
MDL No.: MFCD00037931
Formula:
C₈H₆KNO₄S Melting Pt: 165 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00037931 CAS Number: 2642-37-7 EINECS: 220-145-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...