Methyl-4-chlorocinnamate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-(4-chlorophenyl)acrylate, 4-Chlorocinnamic Acid Methyl Ester
A17071-06
A17071-14
CAAAA17071-03EA
77.14
CAD
CAAAA17071-03
CAAAA17071-06
CAAAA17071-14
Methyl-4-chlorocinnamate 99%
Methyl-4-chlorocinnamate
Purity: 99%
CAS No.: 7560-44-3
Molecular Formula: C10H9ClO2
Formula Weight: 196.64
Melting Point: 74-77°
Beilstein Reference: 2045629
MDL No.: MFCD00075790
Formula:
C₁₀H₉ClO₂ MW: 196.63 g/mol Melting Pt: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075790 CAS Number: 7560-44-3 EINECS: 231-451-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...