4-(1,3-Dithiolan-2-yl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

A17066-06 A17066-14
CAAAA17066-06EA 142.04 CAD
CAAAA17066-06 CAAAA17066-14
4-(1,3-Dithiolan-2-yl)phenol 97%
4-(1,3-Dithiolan-2-yl)phenol

Purity: 97% CAS No.: 22068-49-1 Molecular Formula: C9H10OS2 Formula Weight: 198.30 Melting Point: 117-119° MDL No.: MFCD00068127

Formula: C₉H₁₀OS₂
MW: 198.30 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00068127
CAS Number: 22068-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR