Rose Bengal

Supplier: Thermo Scientific Chemicals

A17053-36 A17053-14 A17053-22
CAAAAA17053-36EA 1020.68 CAD
CAAAAA17053-36 CAAAA17053-14 CAAAA17053-22
Rose Bengal
Rose Bengal

CAS No.: 632-69-9 Molecular Formula: C20H2Cl4I4Na2O5 Formula Weight: 1017.65 Melting Point: 300° Beilstein Reference: 3645857 MDL No.: MFCD00151169

Formula: C₂₀H₄Cl₄I₄O₅
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00151169
CAS Number: 632-69-9
EINECS: 211-183-3
Merck Index: 14,8262

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR