Ethyl-2-thiouracil-5-carboxylate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-Carbethoxy-2-thiouracil
A17046-06
A17046-14
CAAAA17046-06EA
53.25
CAD
CAAAA17046-06
CAAAA17046-14
Ethyl-2-thiouracil-5-carboxylate 99%
Ethyl-2-thiouracil-5-carboxylate
Purity: 99%
CAS No.: 38026-46-9
Molecular Formula: C7H8N2O3S
Formula Weight: 200.22
Melting Point: ca 250° dec.
Beilstein Reference: 190867
MDL No.: MFCD00006074
Formula:
C₇H₈N₂O₃S Melting Pt: ca 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006074 CAS Number: 38026-46-9 EINECS: 253-749-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...