L-Methionine sulfone 99+%

Supplier: Thermo Scientific Chemicals

A17027-03 A17027-06
CAAAA17027-03EA 77.26 CAD
CAAAA17027-03 CAAAA17027-06
L-Methionine sulfone 99+%
L-Methionine sulphone

Purity: 99+% CAS No.: 7314-32-1 Molecular Formula: C5H11NO4S Formula Weight: 181.21 Melting Point: 254° dec. Beilstein Reference: 1725510

Formula: C₅H₁₁NO₄S
Melting Pt: 254 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066020
CAS Number: 7314-32-1
EINECS: 230-774-7

Order Now

Specification Test Results

Optical rotation +11.6° (c=0.69 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR