1-Phenylpiperazine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16995-14
A16995-22
CAAAA16995-14EA
97.17
CAD
CAAAA16995-14
CAAAA16995-22
1-Phenylpiperazine hydrochloride 99%
1-Phenylpiperazine hydrochloride
Purity: 99%
CAS No.: 2210-93-7
Molecular Formula: C10H14N2.HCl
Formula Weight: 198.70
Odor: Odorless
Melting Point: 247-250°
UN No.: 2811
MDL No.: MFCD00012758
Formula:
C₁₀H₁₄N₂.HCl Melting Pt: 247…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012758 CAS Number: 2210-93-7 EINECS: 218-646-9 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...