1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%

Supplier: Thermo Scientific Chemicals
Danger

A16974-06 A16974-14
CAAAA16974-06EA 68.73 CAD
CAAAA16974-06 CAAAA16974-14
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one

Xanthine oxidase inhibitor; decreases uric acid production


Purity: 98% CAS No.: 315-30-0 Molecular Formula: C5H5N4O Formula Weight: 136.11 Odor: Odorless Melting Point: 300° UN No.: 2811 MDL No.: MFCD00599413

Formula: C5H4N4O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00599413
CAS Number: 315-30-0
EINECS: 206-250-9
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR