5-Aminolevulinic acid hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
A16942-04
A16942-ME
CAAAAA16942-04EA
440.46
CAD
CAAAAA16942-04
CAAAA16942-ME
5-Aminolevulinic acid hydrochloride 99%
5-Aminolevulinic acid hydrochloride
Purity: 99%
CAS No.: 5451-09-2
Molecular Formula: C5H9NO3.HCl
Formula Weight: 167.59
Storage Temperature: 2°C to 8°C
Melting Point: ca 156° dec.
Beilstein Reference: 3690651
MDL No.: MFCD00012869
Formula:
C₅H₁₀ClNO₃ MW: 167.59 g/mol Melting Pt: ca 156 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012869 CAS Number: 5451-09-2 EINECS: 226-679-5 Merck Index: 14,446 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...