L(+)-Diisopropyl tartrate 98+%
Supplier: Thermo Scientific Chemicals
A16941-14
A16941-22
CAAAA16941-14EA
78.05
CAD
CAAAA16941-14
CAAAA16941-22
L(+)-Diisopropyl tartrate 98+%
L(+)-Diisopropyl tartrate
Purity: 98+%
CAS No.: 2217-15-4
Molecular Formula: C10H18O6
Formula Weight: 234.25
Flash Point: 110°(230°F)
Refractive Index: 1.4400
Boiling Point: 150-151°/11mm
Beilstein Reference: 1727863
Optical Rotation: +16° (neat)
MDL No.: MFCD00064450
Formula:
C₁₀H₁₈O₆ Boiling Pt: 150…151 °C/11 mm Density: 1.117 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064450 CAS Number: 2217-15-4 EINECS: 218-709-0 |
Specification Test Results
Optical rotation | +16° (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...