2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16922-0F
A16922-AE
A16922-AP
CAAAA16922-0FEA
1409.36
CAD
CAAAA16922-0F
CAAAA16922-AE
CAAAA16922-AP
2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
2-Mercaptopyridine N-oxide sodium salt
CAS No.: 3811-73-2
Molecular Formula: C5H4NNaOS
Formula Weight: 149.15
Refractive Index: 1.4825
Beilstein Reference: 4026050
Merck Reference: 14,7994
MDL No.: MFCD01941547
Formula:
C₅H₄NNaOS Density: 1.238 Storage Temperature: Ambient |
MDL Number:
MFCD01941547 CAS Number: 3811-73-2 EINECS: 223-296-5 UN: 3082 ADR: 9,III Merck Index: 14,7994 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...