Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-amino-4-methylthiazole-5-carboxylate
A16896-06
A16896-14
CAAAA16896-06EA
118.5
CAD
CAAAA16896-06
CAAAA16896-14
CAAAA16896-22
Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Ethyl 2-amino-4-methylthiazole-5-carboxylate
Purity: 97%
CAS No.: 7210-76-6
Molecular Formula: C7H10N2O2S
Formula Weight: 186.22
Odor: Odorless
Melting Point: 172-176°
MDL No.: MFCD00123414
Formula:
C₇H₁₀N₂O₂S MW: 186.22 g/mol Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00123414 CAS Number: 7210-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...