L-Lysine ethyl ester dihydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16824-14
A16824-22
CAAAA16824-14EA
139.99
CAD
CAAAA16824-14
CAAAA16824-22
L-Lysine ethyl ester dihydrochloride 99%
L-Lysine ethyl ester dihydrochloride
Purity: 99%
CAS No.: 3844-53-9
Molecular Formula: C8H18N2O2.2HCl
Formula Weight: 247.17
Melting Point: ca 145° dec.
Beilstein Reference: 3697099
MDL No.: MFCD00039068
Formula:
C₈H₂₀Cl₂N₂O₂ Melting Pt: ca 145 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00039068 CAS Number: 3844-53-9 EINECS: 223-340-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...