L-(-)-Fucose 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(-)-Fucose, 6-Deoxy-L-Galactose, (2S,3R,4R,5S)-2,3,4,5-Tetrahydroxyhexanal
A16789-03
A16789-06
CAAAA16789-03EA
71.26
CAD
CAAAA16789-03
CAAAA16789-06
L-(-)-Fucose 99%
L(-)-Fucose
Purity: 99%
CAS No.: 2438-80-4
Molecular Formula: C6H12O5
Formula Weight: 164.16
Melting Point: 139-142°
Beilstein Reference: 1422661
Merck Reference: 14,4279
Optical Rotation: -75° (c=10 in water)
MDL No.: MFCD00069812
Formula:
C₆H₁₂O₅ MW: 164.16 g/mol Melting Pt: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069812 CAS Number: 2438-80-4 EINECS: 219-452-7 Merck Index: 14,4279 |
Specification Test Results
Optical rotation | -75° (c=10 in water, 24h) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...