Evans blue

Supplier: Thermo Scientific Chemicals
Danger

A16774-09 A16774-18
CAAAA16774-09EA 92.07 CAD
CAAAA16774-09 CAAAA16774-18
Evans blue
Evans blue

Direct Blue 53. Inhibits L-glutamate uptake, and AMPA and kainate receptor currents


CAS No.: 314-13-6 Molecular Formula: C34H24N6Na4O14S4 Formula Weight: 960.82 MDL No.: MFCD00004021


Applications: Direct Blue 53. Inhibits L-glutamate uptake, and AMPA and kainate receptor currents

Formula: C₃₄H₂₈N₆O₁₄S₄
Storage Temperature: Ambient
MDL Number: MFCD00004021
CAS Number: 314-13-6
EINECS: 206-242-5
Merck Index: 14,3905

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR