Phenylmethanesulfonamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A16740-03 A16740-06
CAAAA16740-03EA 91.64 CAD
CAAAA16740-03 CAAAA16740-06
Phenylmethanesulfonamide 98%
Phenylmethanesulfonamide

Purity: 98% CAS No.: 4563-33-1 Molecular Formula: C7H9NO2S Formula Weight: 171.22 Melting Point: 101-104° MDL No.: MFCD00041893

Formula: C7H9NO2S
Melting Pt: 101…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00041893
CAS Number: 4563-33-1
EINECS: 224-935-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR