1,6-Anhydro-β-D-glucose 99%
Supplier: Thermo Scientific Chemicals
A16724-01
A16724-03
CAAAA16724-01EA
63.83
CAD
CAAAA16724-01
CAAAA16724-03
1,6-Anhydro-β-D-glucose 99%
1,6-Anhydro-beta-D-glucose
Purity: 99%
CAS No.: 498-07-7
Molecular Formula: C6H10O5
Formula Weight: 162.14
Melting Point: 180-184°
Beilstein Reference: 80998
MDL No.: MFCD00063248
Formula:
C₆H₁₀O₅ Melting Pt: 180…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063248 CAS Number: 498-07-7 EINECS: 207-855-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...