N-Carbobenzyloxy-L-alanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine
A16716-06
A16716-14
A16716-22
CAAAA16716-06EA
40.92
CAD
CAAAA16716-06
CAAAA16716-14
CAAAA16716-22
N-Carbobenzyloxy-L-alanine 98%
N-Carbobenzyloxy-L-alanine
Purity: 98%
CAS No.: 1142-20-7
Molecular Formula: C11H13NO4
Formula Weight: 223.23
Melting Point: 85-87°
Beilstein Reference: 2056164
Optical Rotation: -14.5° (c=2 in acetic acid)
MDL No.: MFCD00002640
Formula:
C₁₁H₁₃NO₄ Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002640 CAS Number: 1142-20-7 EINECS: 214-532-8 |
Specification Test Results
Optical rotation | -14.5° (c=2 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...