3-Noradamantanecarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16683-01
A16683-03
A16683-06
CAAAA16683-01EA
44.71
CAD
CAAAA16683-01
CAAAA16683-03
CAAAA16683-06
3-Noradamantanecarboxylic acid 98%
3-Noradamantanecarboxylic acid
Purity: 98%
CAS No.: 16200-53-6
Molecular Formula: C10H14O2
Formula Weight: 166.22
Melting Point: 105-107°
MDL No.: MFCD00213463
Formula:
C₁₀H₁₄O₂ Melting Pt: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213463 CAS Number: 16200-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...