2,5-Dithiobiurea 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydrazine-1,2-bis(carbothioamide)

A16667-14 A16667-22
CAAAA16667-14EA 63.04 CAD
CAAAA16667-14 CAAAA16667-22
2,5-Dithiobiurea 97%
2,5-Dithiobiurea

Purity: 97% CAS No.: 142-46-1 Molecular Formula: C2H6N4S2 Formula Weight: 150.22 Melting Point: ca 212° dec. Beilstein Reference: 1723178 MDL No.: MFCD00014471

Formula: C₂H₆N₄S₂
MW: 150.22 g/mol
Melting Pt: ∼ 212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014471
CAS Number: 142-46-1
EINECS: 205-537-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR