Methyl-2-amino-5-bromobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16666-06
A16666-14
A16666-22
CAAAA16666-06EA
58.46
CAD
CAAAA16666-06
CAAAA16666-14
CAAAA16666-22
Methyl-2-amino-5-bromobenzoate 98%
Methyl 2-amino-5-bromobenzoate
Purity: 98%
CAS No.: 52727-57-8
Molecular Formula: C8H8BrNO2
Formula Weight: 230.06
Melting Point: 71-73°
Beilstein Reference: 972720
Formula:
C₈H₈BrNO₂ Melting Pt: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00144761 CAS Number: 52727-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...