1-Cyclohexyl-2-pyrrolidone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16638-18
A16638-30
CAAAA16638-18EA
41.87
CAD
CAAAA16638-18
CAAAA16638-30
1-Cyclohexyl-2-pyrrolidone 99%
1-Cyclohexyl-2-pyrrolidone
Purity: 99%
CAS No.: 6837-24-7
Molecular Formula: C10H17NO
Formula Weight: 167.25
Flash Point: 140°(284°F)
Refractive Index: 1.4990
Melting Point: 12-15°
Boiling Point: 290-292°
Beilstein Reference: 121832
UN No.: 2810
MDL No.: MFCD00003191
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Boiling Pt: 290…292 °C Melting Pt: 12…15 °C Density: 1.007 Flash Pt: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003191 CAS Number: 6837-24-7 EINECS: 229-919-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...