4-(Dimethylamino)butanoic acid hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16610-06
A16610-14
CAAAA16610-06EA
94.33
CAD
CAAAA16610-06
CAAAA16610-14
4-(Dimethylamino)butanoic acid hydrochloride 98%
4-(Dimethylamino)butanoic acid hydrochloride
Purity: 98%
CAS No.: 69954-66-1
Molecular Formula: HO2C(CH2)3N(CH3)2.HCl
Formula Weight: 167.64
Melting Point: 153-155°
MDL No.: MFCD00012613
Formula:
C₆H₁₄ClNO₂ Melting Pt: 153…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012613 CAS Number: 69954-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...