Chromium(III)-2-ethylhexanoate 50% in 2-ethylhexanoic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chromium(III) 2-ethylhexanoate

A16560-18 A16560-30
CAAAAA16560-18EA 137.46 CAD
CAAAAA16560-18 CAAAAA16560-30
Chromium(III)-2-ethylhexanoate 50% in 2-ethylhexanoic acid
Chromium(III)-2-ethylhexanoate

CAS No.: 3444-17-5 Molecular Formula: C24H45CrO6 Formula Weight: 481.62 Odor: Weak, characteristic MDL No.: MFCD00014005

Formula: C₂₄H₄₅CrO₆
MW: 481.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014005
CAS Number: 3444-17-5
EINECS: 222-357-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR