7-Hydroxy-4-(trifluoromethyl)coumarin 98%

Supplier: Thermo Scientific Chemicals
Warning

A16537-03 A16537-06
CAAAA16537-03EA 111.39 CAD
CAAAA16537-03 CAAAA16537-06
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
7-Hydroxy-4-(trifluoromethyl)coumarin

Purity: 98% CAS No.: 575-03-1 Molecular Formula: C10H5F3O3 Formula Weight: 230.14 Melting Point: 178-180° MDL No.: MFCD00037578

Formula: C₁₀H₅F₃O₃
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00037578
CAS Number: 575-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR