3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

A16317-06 A16317-14 A16317-22
CAAAAA16317-06EA 81.21 CAD
CAAAAA16317-06 CAAAAA16317-14 CAAAAA16317-22
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate 97%
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate

Purity: 97% CAS No.: 149022-15-1 Molecular Formula: C8H9N3S.HCl.xH2O Formula Weight: 215.71(anhy) Melting Point: ca 275° dec. Beilstein Reference: 3571041 UN No.: 2811 MDL No.: MFCD00149370

Formula: C₈H₉N₃S
Melting Pt: ca 275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149370
CAS Number: 149022-15-1
EINECS: 238-428-7
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR