(Methoxycarbonylmethyl)triphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Methoxy-2-oxoethyl)triphenylphosphonium bromide

A16301-14 A16301-22 A16301-36
CAAAA16301-14EA 58.93 CAD
CAAAA16301-14 CAAAA16301-22 CAAAA16301-36
(Methoxycarbonylmethyl)triphenylphosphonium bromide 98+%
(Methoxycarbonylmethyl)triphenylphosphonium bromide

Purity: 98+% CAS No.: 1779-58-4 Molecular Formula: CH3O2CCH2P(C6H5)3Br Formula Weight: 415.28 Melting Point: ca 163° dec. Beilstein Reference: 3812975

Formula: C₂₁H₂₀BrO₂P
MW: 415.28 g/mol
Melting Pt: ∼ 163 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011801
CAS Number: 1779-58-4
EINECS: 217-222-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR