Calcium glycerophosphate hydrate 97%
Supplier: Thermo Scientific Chemicals
A16269-0B
A16269-30
CAAAA16269-0BEA
320.74
CAD
CAAAA16269-0B
CAAAA16269-30
Calcium glycerophosphate hydrate 97%
Calcium glycerophosphate hydrate
Purity: 97%
CAS No.: 28917-82-0
Molecular Formula: C3H7CaO6P.xH2O
Formula Weight: 210.15(anhy)
MDL No.: MFCD00151656
Formula:
C₃H₇CaO₆P·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00151656 CAS Number: 28917-82-0 EINECS: 249-312-0 Merck Index: 14,1670 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...