N-Boc-L-serine, (max. 10% H₂O) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate

A16224-06 A16224-14
CAAAA16224-03EA 27.87 CAD
CAAAA16224-03 CAAAA16224-06 CAAAA16224-14
N-Boc-L-serine, (max. 10% H₂O) 98%
N-Boc-L-serine

Purity: 98% CAS No.: 3262-72-4 Molecular Formula: C8H15NO5 Formula Weight: 205.21 Storage Temperature: 2°C to 8°C Melting Point: 90-91° dec. Beilstein Reference: 2212252 Optical Rotation: -3.5° (c=2 in acetic acid) MDL No.: MFCD00037243

Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
Melting Pt: 90…91 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00037243
CAS Number: 3262-72-4
EINECS: 221-867-3

Order Now

Specification Test Results

Optical rotation -3.5° (c=2 in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR